accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOOONOOHOOHNH4NH4H2O

CAS RN 304675-80-7

IUPAC Name: diammonium 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxylatomethyl)amino)acetate hydrate
Synonyms: diammonium 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxylatomethyl)amino)acetate hydrate
Molecular Weight (g/mol): 344.32
Molecular Formula: C10H24N4O9
InChi Key: WRSUMHYBOYWMTD-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].O.OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results