accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3COCl

CAS RN 304685-85-6

IUPAC Name: 2-chloro-1-(2,5-dimethyl-1-phenyl-1H-pyrrol-3-yl)ethan-1-one
Synonyms: 2-chloro-1-(2,5-dimethyl-1-phenylpyrrol-3-yl)ethanone
Molecular Weight (g/mol): 247.72
Molecular Formula: C14H14ClNO
InChi Key: QSYYGYUKZQOGRQ-UHFFFAOYSA-N
SMILES: CC1=CC(C(=O)CCl)=C(C)N1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results