accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 304851-95-4

IUPAC Name: yttrium(3+) tritrifluoroacetate
Synonyms: yttrium(3+) tritrifluoroacetate
Molecular Weight (g/mol): 427.95
Molecular Formula: C6F9O6Y
InChi Key: NPATVYPXYMSPKD-UHFFFAOYSA-K
SMILES: [Y+3].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results