accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNOOClClN(E/Z)

CAS RN 304896-28-4

IUPAC Name: 2-cyano-3-[5-(2,5-dichlorophenyl)furan-2-yl]-N-(quinolin-5-yl)prop-2-enamide
Synonyms: 2-cyano-3-[5-(2,5-dichlorophenyl)furan-2-yl]-N-(quinolin-5-yl)prop-2-enamide
Molecular Weight (g/mol): 434.28
Molecular Formula: C23H13Cl2N3O2
InChi Key: SVENPFFEMUOOGK-UHFFFAOYSA-N
SMILES: ClC1=CC(C2=CC=C(O2)C=C(C#N)C(=O)NC2=C3C=CC=NC3=CC=C2)=C(Cl)C=C1
Product Line 
  • (2)
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results