accessibility menu, dialog, popup

UserName

Viewing profile as user:
OPOO

CAS RN 3049-24-9

IUPAC Name: diphenyl phenylphosphonate
Synonyms: diphenyl phenylphosphonate
Molecular Weight (g/mol): 310.29
Molecular Formula: C18H15O3P
InChi Key: CDOMXXVCZQOOMT-UHFFFAOYSA-N
SMILES: O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results