accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30491-74-8

IUPAC Name: 5-(4-aminophenoxy)-2-nitroaniline
Synonyms: 5-(4-aminophenoxy)-2-nitroaniline
Molecular Weight (g/mol): 245.24
Molecular Formula: C12H11N3O3
InChi Key: ZHELWQKSRPWTPN-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC2=CC(N)=C(C=C2)[N+]([O-])=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results