CAS RN 3050-69-9
CAS RN 3050-69-9
IUPAC Name:
ethenyl hexanoate
Synonyms:
ethenyl hexanoate
Molecular Weight (g/mol):
142.2
Molecular Formula:
C8H14O2
InChi Key:
LZWYWAIOTBEZFN-UHFFFAOYSA-N
SMILES:
CCCCCC(=O)OC=C
1
–
1
of
1
results
1
Vinyl Hexanoate (stabilized with MEHQ) 99.0+%, TCI America™

CAS: 3050-69-9 Molecular Formula: C8H14O2 Molecular Weight (g/mol): 142.20 MDL Number: MFCD00048869 InChI Key: LZWYWAIOTBEZFN-UHFFFAOYSA-N Synonym: Hexanoic Acid Vinyl Ester PubChem CID: 76451 IUPAC Name: ethenyl hexanoate SMILES: CCCCCC(=O)OC=C