CAS RN 3053-68-7
IUPAC Name:
triphenylborane; triphenylphosphane
Synonyms:
triphenylborane; triphenylphosphine
Molecular Weight (g/mol):
504.42
Molecular Formula:
C36H30BP
InChi Key:
QLAGHGSFXJZWKY-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)B(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Triphenylborane - Triphenylphosphine Complex 97.0+%, TCI America™
CAS: 3053-68-7 Molecular Formula: C36H30BP Molecular Weight (g/mol): 504.42 MDL Number: MFCD01631317 InChI Key: QLAGHGSFXJZWKY-UHFFFAOYSA-N PubChem CID: 11081676 IUPAC Name: triphenylborane;triphenylphosphane SMILES: B(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3