accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3

CAS RN 3056-59-5

IUPAC Name: 2-ethylphenyl acetate
Synonyms: 2-ethylphenyl acetate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: VERVERDPMMBDSS-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1OC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results