accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOHNH3COSH3C

CAS RN 30560-19-1

IUPAC Name: N-[methoxy(methylsulfanyl)phosphoryl]acetamide
Synonyms: acephate acetamidophos
Molecular Weight (g/mol): 183.16
Molecular Formula: C4H10NO3PS
InChi Key: YASYVMFAVPKPKE-UHFFFAOYNA-N
SMILES: COP(=O)(NC(C)=O)SC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results