accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2NNH3COSOOO

CAS RN 30578-37-1

IUPAC Name: 4-amino-6-methoxy-1-phenyl-1λ⁵-pyridazin-1-ylium methyl sulfate
Synonyms: amezinium metilsulfate 甲硫阿美铵
Molecular Weight (g/mol): 313.33
Molecular Formula: C12H15N3O5S
InChi Key: ZEASXVYVFFXULL-UHFFFAOYSA-N
SMILES: COS([O-])(=O)=O.COC1=CC(N)=CN=[N+]1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results