accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3058-04-6

IUPAC Name: 3-[9-(2-cyanoethyl)-2,4,8,10-tetraoxaspiro[5.5]undecan-3-yl]propanenitrile
Synonyms: 3-[9-(2-cyanoethyl)-2,4,8,10-tetraoxaspiro[5.5]undecan-3-yl]propanenitrile
Molecular Weight (g/mol): 266.3
Molecular Formula: C13H18N2O4
InChi Key: BNAMIPLIODPZLE-UHFFFAOYSA-N
SMILES: N#CCCC1OCC2(CO1)COC(CCC#N)OC2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results