accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 305800-58-2

IUPAC Name: 2-(3-bromo-5-fluorophenyl)acetonitrile
Synonyms: 2-(3-bromo-5-fluorophenyl)acetonitrile
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H5BrFN
InChi Key: FSKMLBLIAOVQCF-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(CC#N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results