accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONCH3OHNBr

CAS RN 3060-89-7

IUPAC Name: 1-(4-bromophenyl)-3-methoxy-3-methylurea
Synonyms: metobromuron
Molecular Weight (g/mol): 259.1
Molecular Formula: C9H11BrN2O2
InChi Key: WLFDQEVORAMCIM-UHFFFAOYSA-N
SMILES: CON(C)C(=O)NC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results