accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHHOOOOHHOOOOHHOH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OErEr

CAS RN 30618-31-6

IUPAC Name: tris(oxalic acid) dierbium decahydrate
Synonyms: tris(oxalic acid) dierbium decahydrate
Molecular Weight (g/mol): 784.77
Molecular Formula: C6H26Er2O22
InChi Key: CYHKKMLSGVLNPD-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.O.O.O.O.[Er].[Er].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)