accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3065-79-0

IUPAC Name: 3-ethyl-2-{3-[(2Z)-3-ethyl-2,3-dihydro-1,3-benzothiazol-2-ylidene]-2-methylprop-1-en-1-yl}-1,3-benzothiazol-3-ium iodide
Synonyms: 3-ethyl-2-{3-[(2Z)-3-ethyl-1,3-benzothiazol-2-ylidene]-2-methylprop-1-en-1-yl}-1,3-benzothiazol-3-ium iodide
Molecular Weight (g/mol): 506.46
Molecular Formula: C22H23IN2S2
InChi Key: SAEMBGFHGROQJZ-UHFFFAOYSA-M
SMILES: [I-].CCN1\C(SC2=CC=CC=C12)=C\C(C)=CC1=[N+](CC)C2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results