accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOOHO

CAS RN 30651-24-2

IUPAC Name: 5-nitropyridine-2-carboxylic acid
Synonyms: 5-nitropyridine-2-carboxylic acid
Molecular Weight (g/mol): 168.11
Molecular Formula: C6H4N2O4
InChi Key: QKYRCTVBMNXTBT-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)