accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOH3CHO

CAS RN 30652-11-0

IUPAC Name: 3-hydroxy-1,2-dimethyl-1,4-dihydropyridin-4-one
Synonyms: ferriprox dmohpo deferiprone (product) chiesi brand of deferiprone deferiprone L1 oral chelate Hdpp l1 oral chelate CP20 (chelating agent) hdmpp cpd
Molecular Weight (g/mol): 139.15
Molecular Formula: C7H9NO2
InChi Key: TZXKOCQBRNJULO-UHFFFAOYSA-N
SMILES: CN1C=CC(=O)C(O)=C1C
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)