CAS RN 30675-13-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ UCH-L3 Inhibitor
The UCH-L3 Inhibitor, also referenced under CAS 30675-13-9, controls the biological activity of UCH-L3. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.