CAS RN 306769-48-2
IUPAC Name:
2-bromo-3-dodecyl-5-iodothiophene
Synonyms:
2-bromo-3-dodecyl-5-iodothiophene
Molecular Weight (g/mol):
457.25
Molecular Formula:
C16H26BrIS
InChi Key:
NUMNAXSVZXXUAM-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCC1=C(Br)SC(I)=C1
1
–
1
of
1
results
1
2-Bromo-3-dodecyl-5-iodothiophene (stabilized with Copper chip) 95.0+%, TCI America™
CAS: 306769-48-2 Molecular Formula: C16H26BrIS Molecular Weight (g/mol): 457.252 InChI Key: NUMNAXSVZXXUAM-UHFFFAOYSA-N PubChem CID: 91972091 IUPAC Name: 2-bromo-3-dodecyl-5-iodothiophene SMILES: CCCCCCCCCCCCC1=C(SC(=C1)I)Br