accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30684-06-1

IUPAC Name: 4-phenylbutan-1-amine; methane
Synonyms: 4-phenylbutylamine; methane
Molecular Weight (g/mol): 165.28
Molecular Formula: C11H19N
InChi Key: VGECFYQRUARHKB-UHFFFAOYSA-N
SMILES: C.NCCCCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results