accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSBrNH2

CAS RN 306934-99-6

IUPAC Name: ethyl 2-amino-4-(4-bromophenyl)thiophene-3-carboxylate
Synonyms: ethyl 2-amino-4-(4-bromophenyl)thiophene-3-carboxylate
Molecular Weight (g/mol): 326.21
Molecular Formula: C13H12BrNO2S
InChi Key: SEWFWRCESBYGFS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results