accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSOBr

CAS RN 306935-06-8

IUPAC Name: 2-bromo-1-[5-(pyridin-2-yl)thiophen-2-yl]ethan-1-one
Synonyms: 2-bromo-1-[5-(pyridin-2-yl)thiophen-2-yl]ethanone
Molecular Weight (g/mol): 282.16
Molecular Formula: C11H8BrNOS
InChi Key: MJDNWZQQRFCXRU-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C(S1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results