CAS RN 306974-70-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ GPR40 Antagonist, GW1100
The GPR40 Antagonist, GW1100, also referenced under CAS 306974-70-9, controls the biological activity of GPR40. This small molecule/inhibitor is primarily used for Biochemicals applications.