accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NCH3CH3H3CNCH3NCH3CH3H2OH2OClCuClCu

CAS RN 30698-64-7

IUPAC Name: bis([2-(dimethylamino)ethyl]dimethylamine) bis(chlorocopper) dihydrate
Synonyms: bis(cuprous chloride) bis(temed) dihydrate
Molecular Weight (g/mol): 466.44
Molecular Formula: C12H36Cl2Cu2N4O2
InChi Key: VVXKYYDFGPZSOZ-UHFFFAOYSA-L
SMILES: O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)