accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 307-78-8

IUPAC Name: hexadecafluorodecanedioic acid
Synonyms: hexadecafluorodecanedioic acid
Molecular Weight (g/mol): 490.1
Molecular Formula: C10H2F16O4
InChi Key: YPCSMEGZIYWAAZ-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results