accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFFFFFFFFFFFFFFFFFFF

CAS RN 307-79-9

IUPAC Name: methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoate
Synonyms: methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoate
Molecular Weight (g/mol): 528.11
Molecular Formula: C11H3F19O2
InChi Key: QJFHNYDPNSFJMR-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results