accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFFFFFFFOOH2C

CAS RN 307-98-2

IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl prop-2-enoate
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl prop-2-enoate
Molecular Weight (g/mol): 454.14
Molecular Formula: C11H5F15O2
InChi Key: YSQGYEYXKXGAQA-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)COC(=O)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results