accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3073-05-0

IUPAC Name: 4-{[1,1'-biphenyl]-4-yl}-4'-phenyl-1,1'-biphenyl
Synonyms: 4-{[1,1'-biphenyl]-4-yl}-4'-phenyl-1,1'-biphenyl
Molecular Weight (g/mol): 382.51
Molecular Formula: C30H22
InChi Key: OMCUOJTVNIHQTI-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results