accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3073-59-4

H3CONHHNCH3O

CAS RN 3073-59-4

IUPAC Name: N-(6-acetamidohexyl)acetamide
Synonyms: N-(6-acetamidohexyl)acetamide
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: BNQSTAOJRULKNX-UHFFFAOYSA-N
SMILES: CC(=O)NCCCCCCNC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)