accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3074-03-1

IUPAC Name: 11H-benzo[b]fluoren-11-one
Synonyms: benzo[b]fluoren-11-one
Molecular Weight (g/mol): 230.27
Molecular Formula: C17H10O
InChi Key: MLMNDNOSVOKYMT-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C2=C1C=C1C=CC=CC1=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results