accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH2

CAS RN 3074-78-0

IUPAC Name: 2,6-dimethylhept-1-ene
Synonyms: 2,6-dimethylhept-1-ene
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: JUUMAEXLYIMEOD-UHFFFAOYSA-N
SMILES: CC(C)CCCC(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results