accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30748-47-1

IUPAC Name: 1-(2-amino-4-methyl-1,3-thiazol-5-yl)ethan-1-one
Synonyms: 1-(2-amino-4-methyl-1,3-thiazol-5-yl)ethanone
Molecular Weight (g/mol): 156.2
Molecular Formula: C6H8N2OS
InChi Key: PKUKCASRNJIQNU-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)N=C(N)S1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)