accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2FFZnBr

CAS RN 307496-26-0

IUPAC Name: (2,4-difluorophenyl)methanide; bromozincylium
Synonyms: (2,4-difluorophenyl)methanide; bromozincylium
Molecular Weight (g/mol): 272.4
Molecular Formula: C7H5BrF2Zn
InChi Key: WPJAHFYOACWPNA-UHFFFAOYSA-M
SMILES: [Zn+]Br.[CH2-]C1=C(F)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results