CAS RN 30751-05-4
CAS RN 30751-05-4
IUPAC Name:
3,4,5-trimethoxy-N-(piperidin-3-yl)benzamide
Synonyms:
troxipide
Molecular Weight (g/mol):
294.35
Molecular Formula:
C15H22N2O4
InChi Key:
YSIITVVESCNIPR-UHFFFAOYNA-N
SMILES:
COC1=CC(=CC(OC)=C1OC)C(=O)NC1CCCNC1