accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBr

CAS RN 30752-19-3

IUPAC Name: 1-bromo-4-(hexyloxy)benzene
Synonyms: 1-bromo-4-(hexyloxy)benzene
Molecular Weight (g/mol): 257.17
Molecular Formula: C12H17BrO
InChi Key: GKLMJONYGGTHHM-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)