accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3077-46-1

IUPAC Name: (2S,3S)-2-acetamido-3-methylpentanoic acid
Synonyms: N-acetylisoleucine
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: JDTWZSUNGHMMJM-IFSCUNPJNA-N
SMILES: CC[C@H](C)[C@H](NC(C)=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)