accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30838-93-8

IUPAC Name: 2-chloro-4,6-dimethylpyridine
Synonyms: 2-chloro-4,6-dimethylpyridine
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: COVGXNSRDOYAJD-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results