accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOHOOH3C

CAS RN 3084-40-0

IUPAC Name: diethyl (hydroxymethyl)phosphonate
Synonyms: diethyl hydroxymethylphosphonate
Molecular Weight (g/mol): 168.13
Molecular Formula: C5H13O4P
InChi Key: RWIGWWBLTJLKMK-UHFFFAOYSA-N
SMILES: CCOP(=O)(CO)OCC
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)