accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3086-62-2

IUPAC Name: 3,4,5-trimethoxybenzamide
Synonyms: 3,4,5-trimethoxybenzamide
Molecular Weight (g/mol): 211.22
Molecular Formula: C10H13NO4
InChi Key: GGNMTJKRHHLJHH-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1OC)C(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results