accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3087-16-9

IUPAC Name: sodium bis[4-(dimethylamino)phenyl](2-hydroxy-3,6-disulfonatonaphthalen-1-yl)methylium
Synonyms: acid green 50
Molecular Weight (g/mol): 576.61
Molecular Formula: C27H25N2NaO7S2
InChi Key: FVZFYEQFFLFVJA-UHFFFAOYSA-M
SMILES: [Na+].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=C2C=CC(=CC2=CC(=C1O)S([O-])(=O)=O)S([O-])(=O)=O
Grade 
  • (1)
  • (1)
  • (1)
Quantity 
  • (2)
Color 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)