accessibility menu, dialog, popup

UserName

Viewing profile as user:
TiH3CCH3CH3OCH3CH3CH3OCH3CH3CH3OCH3CH3CH3O

CAS RN 3087-39-6

IUPAC Name: titanium(4+) tetrakis(2-methylpropan-2-olate)
Synonyms: titanum tetra T-butoxide
Molecular Weight (g/mol): 340.33
Molecular Formula: C16H36O4Ti
InChi Key: GRWPYGBKJYICOO-UHFFFAOYSA-N
SMILES: [Ti+4].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-]
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)