CAS RN 308795-98-4
IUPAC Name:
zinc(2+) 6-methylpyridin-2-ide bromide
Synonyms:
zinc(2+) 6-methylpyridin-2-ide bromide
Molecular Weight (g/mol):
237.41
Molecular Formula:
C6H6BrNZn
InChi Key:
JSTYQZIXEXOPBT-UHFFFAOYSA-M
SMILES:
[Zn++].[Br-].CC1=N[C-]=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich 3-Methoxybenzylzinc Chloride Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.