accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 308815-83-0

IUPAC Name: 2-(4-{[(tert-butyldimethylsilyl)oxy]methyl}-2-nitrophenoxy)acetic acid
Synonyms: 4-{[(tert-butyldimethylsilyl)oxy]methyl}-2-nitrophenoxyacetic acid
Molecular Weight (g/mol): 341.44
Molecular Formula: C15H23NO6Si
InChi Key: UXUZQGZIXUXYHB-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OCC1=CC(=C(OCC(O)=O)C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results