accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClSnH

CAS RN 3091-32-5

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 405.64
Molecular Formula: C18H35ClSn
InChi Key: WLSJDIYSPLULMS-UHFFFAOYSA-N
SMILES: Cl.C1CCC(CC1)[SnH](C1CCCCC1)C1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)