accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CO(R)(S)(S)O(S)H3C(R)F(S)OH(S)H3C(S)OOCl

CAS RN 3093-35-4

IUPAC Name: (1S,2S,4R,8S,9S,11S,12R,13S)-8-(2-chloroacetyl)-12-fluoro-11-hydroxy-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icos-17-en-16-one
Synonyms: halcinonide
Molecular Weight (g/mol): 454.96
Molecular Formula: C24H32ClFO5
InChi Key: MUQNGPZZQDCDFT-JNQJZLCISA-N
SMILES: CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)CC[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results