accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3093-88-7

IUPAC Name: 3-fluorothiophene
Synonyms: 3-fluorothiophene
Molecular Weight (g/mol): 102.13
Molecular Formula: C4H3FS
InChi Key: WPAQIMRFMFRJTP-UHFFFAOYSA-N
SMILES: FC1=CSC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results