CAS RN 3095-38-3
IUPAC Name:
3,5-dimethyl-4-nitrobenzoic acid
Synonyms:
3,5-dimethyl-4-nitrobenzoic acid
Molecular Weight (g/mol):
195.17
Molecular Formula:
C9H9NO4
InChi Key:
RBAVFNOGEPCOQI-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC(C)=C1[N+]([O-])=O)C(O)=O
1
–
1
of
1
results
1
Promotions Available
3,5-Dimethyl-4-nitrobenzoic acid, 97%
3,5-Dimethyl-4-nitrobenzoic acid, 97%
CAS: 3095-38-3 Molecular Formula: C9H9NO4 Molecular Weight (g/mol): 195.17 MDL Number: MFCD00014699 InChI Key: RBAVFNOGEPCOQI-UHFFFAOYSA-N PubChem CID: 249413 IUPAC Name: 3,5-dimethyl-4-nitrobenzoic acid SMILES: CC1=CC(=CC(C)=C1[N+]([O-])=O)C(O)=O