accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBrBrBrBrBr

CAS RN 3095-73-6

IUPAC Name: hexakis(bromomethyl)benzene
Synonyms: hexakis(bromomethyl)benzene
Molecular Weight (g/mol): 635.65
Molecular Formula: C12H12Br6
InChi Key: XJOUCILNLRXRTF-UHFFFAOYSA-N
SMILES: BrCC1=C(CBr)C(CBr)=C(CBr)C(CBr)=C1CBr
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)