accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NO

CAS RN 3096-57-9

IUPAC Name: 2-amino-9H-fluoren-9-one
Synonyms: 2-amino-9-fluorenone
Molecular Weight (g/mol): 195.22
Molecular Formula: C13H9NO
InChi Key: SJODITPGMMSNRF-UHFFFAOYSA-N
SMILES: NC1=CC=C2C3=CC=CC=C3C(=O)C2=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)